Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATATCGGTCCTGGAGAAGACGCTA[C/T]TGTCCATGTCGTCCTGCACAGACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 167780 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102724183 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC102724183 - Wnt family member 9-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107546764 - uncharacterized LOC107546764 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPY - pancreatic polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319209.1 | Intron | NP_001306138.1 | ||||
NM_002722.4 | Intron | NP_002713.1 | ||||
XM_011524978.2 | Intron | XP_011523280.1 |
Set Membership: |
HapMap Validated |