Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604384 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP2C1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATP2C1 - ATPase secretory pathway Ca2+ transporting 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001485.2 | Intron | NP_001001485.1 | ||||
NM_001001486.1 | Intron | NP_001001486.1 | ||||
NM_001001487.1 | Intron | NP_001001487.1 | ||||
NM_001199179.1 | Intron | NP_001186108.1 | ||||
NM_001199180.1 | Intron | NP_001186109.1 | ||||
NM_001199181.1 | Intron | NP_001186110.1 | ||||
NM_001199182.1 | Intron | NP_001186111.1 | ||||
NM_001199183.1 | Intron | NP_001186112.1 | ||||
NM_001199184.1 | Intron | NP_001186113.1 | ||||
NM_001199185.1 | Intron | NP_001186114.1 | ||||
NM_014382.3 | Intron | NP_055197.2 | ||||
XM_005247354.2 | Intron | XP_005247411.1 | ||||
XM_005247355.2 | Intron | XP_005247412.1 | ||||
XM_005247356.2 | Intron | XP_005247413.1 | ||||
XM_005247358.2 | Intron | XP_005247415.1 | ||||
XM_011512686.2 | Intron | XP_011510988.1 | ||||
XM_017006164.1 | Intron | XP_016861653.1 |
Set Membership: |
HapMap |