Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600364 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GUCA1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GUCA1A - guanylate cyclase activator 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000409.4 | Intron | NP_000400.2 | ||||
NM_001319061.1 | Intron | NP_001305990.1 | ||||
NM_001319062.1 | Intron | NP_001305991.1 | ||||
XM_006715073.3 | Intron | XP_006715136.1 | ||||
XM_011514537.2 | Intron | XP_011512839.1 | ||||
XM_011514539.1 | Intron | XP_011512841.1 |