Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606625 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLAMF7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLAMF7 - SLAM family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282588.1 | Intron | NP_001269517.1 | ||||
NM_001282589.1 | Intron | NP_001269518.1 | ||||
NM_001282590.1 | Intron | NP_001269519.1 | ||||
NM_001282591.1 | Intron | NP_001269520.1 | ||||
NM_001282592.1 | Intron | NP_001269521.1 | ||||
NM_001282593.1 | Intron | NP_001269522.1 | ||||
NM_001282594.1 | Intron | NP_001269523.1 | ||||
NM_001282595.1 | Intron | NP_001269524.1 | ||||
NM_001282596.1 | Intron | NP_001269525.1 | ||||
NM_021181.4 | Intron | NP_067004.3 | ||||
XM_011509828.1 | Intron | XP_011508130.1 | ||||
XM_011509829.1 | Intron | XP_011508131.1 |
Set Membership: |
JSNP Validated |