Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAATTTCCAAGCGCCAATCTTCAC[C/G]TCCCGGACTCTAACTCTGCGGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604674 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEY2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HEY2 - hes related family bHLH transcription factor with YRPW motif 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012259.2 | Intron | NP_036391.1 | ||||
XM_017010627.1 | Intron | XP_016866116.1 | ||||
XM_017010628.1 | Intron | XP_016866117.1 | ||||
XM_017010629.1 | Intron | XP_016866118.1 |
LOC105377986 - uncharacterized LOC105377986 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |