Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGCGAGGCGGCATCAGCCCCCC[C/T]AGTTCTCACCCCCCATTAGCAGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609208 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KAZALD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KAZALD1 - Kazal type serine peptidase inhibitor domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319303.1 | Intron | NP_001306232.1 | ||||
NM_030929.4 | Intron | NP_112191.2 | ||||
XM_005270194.3 | Intron | XP_005270251.1 | ||||
XM_017016714.1 | Intron | XP_016872203.1 | ||||
XM_017016715.1 | Intron | XP_016872204.1 |