Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGACACTTTTCTAAGCCTTTTAC[A/C]TACTGTCTCTCAGCAACCACCCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610454 MIM: 612971 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LZTS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LZTS2 - leucine zipper tumor suppressor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDZD7 - PDZ domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195263.1 | Intron | NP_001182192.1 | ||||
NM_024895.4 | Intron | NP_079171.1 | ||||
XM_005270165.2 | Intron | XP_005270222.1 | ||||
XM_011540177.2 | Intron | XP_011538479.1 | ||||
XM_011540178.2 | Intron | XP_011538480.1 | ||||
XM_011540179.2 | Intron | XP_011538481.1 | ||||
XM_011540181.2 | Intron | XP_011538483.1 | ||||
XM_011540182.2 | Intron | XP_011538484.1 | ||||
XM_011540183.2 | Intron | XP_011538485.1 | ||||
XM_017016667.1 | Intron | XP_016872156.1 | ||||
XM_017016668.1 | Intron | XP_016872157.1 |
Set Membership: |
HapMap |