Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606075 MIM: 610454 MIM: 612971 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C10orf2 - chromosome 10 open reading frame 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LZTS2 - leucine zipper tumor suppressor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318099.1 | 2404 | Intron | NP_001305028.1 | |||
NM_001318100.1 | 2404 | Intron | NP_001305029.1 | |||
NM_001318101.1 | 2404 | Intron | NP_001305030.1 | |||
NM_032429.3 | 2404 | Intron | NP_115805.1 | |||
XM_005270222.4 | 2404 | Intron | XP_005270279.1 | |||
XM_017016780.1 | 2404 | UTR 5 | XP_016872269.1 | |||
XM_017016781.1 | 2404 | Intron | XP_016872270.1 |
PDZD7 - PDZ domain containing 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |