Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGCTTCTCCTGCTGAAAGGAGAG[A/G]GGCCCATCACCAGAGCCCCGGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614566 MIM: 602510 MIM: 600782 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAAF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNAAF3 - dynein axonemal assembly factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTPRH - protein tyrosine phosphatase, receptor type H | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYT5 - synaptotagmin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297774.1 | Intron | NP_001284703.1 | ||||
NM_003180.2 | Intron | NP_003171.2 | ||||
XM_006723339.3 | Intron | XP_006723402.1 | ||||
XM_006723340.3 | Intron | XP_006723403.1 | ||||
XM_006723341.3 | Intron | XP_006723404.1 | ||||
XM_017027175.1 | Intron | XP_016882664.1 | ||||
XM_017027176.1 | Intron | XP_016882665.1 |
Set Membership: |
HapMap |