Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCCTGCAGCCTGTGCTCAGTGAG[C/G]TGCTGCACCCAGCTGTTCAAGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC155 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC155 - coiled-coil domain containing 155 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144688.4 | 19 | Intron | NP_653289.3 | |||
XM_005258549.3 | 19 | Intron | XP_005258606.1 | |||
XM_011526488.2 | 19 | Intron | XP_011524790.1 | |||
XM_011526489.2 | 19 | Intron | XP_011524791.1 | |||
XM_011526490.2 | 19 | Intron | XP_011524792.1 | |||
XM_011526494.2 | 19 | Intron | XP_011524796.1 | |||
XM_011526495.2 | 19 | Intron | XP_011524797.1 | |||
XM_011526496.2 | 19 | Intron | XP_011524798.1 | |||
XM_011526497.2 | 19 | Intron | XP_011524799.1 | |||
XM_017026333.1 | 19 | UTR 5 | XP_016881822.1 |
LOC101928295 - uncharacterized LOC101928295 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |