Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGTGCAAATTTCACTCACTGCA[A/G]AGAATCCTGGAAGAGTTAGAGGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC155 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CCDC155 - coiled-coil domain containing 155 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144688.4 | Intron | NP_653289.3 | ||||
XM_005258549.3 | Intron | XP_005258606.1 | ||||
XM_011526488.2 | Intron | XP_011524790.1 | ||||
XM_011526489.2 | Intron | XP_011524791.1 | ||||
XM_011526490.2 | Intron | XP_011524792.1 | ||||
XM_011526494.2 | Intron | XP_011524796.1 | ||||
XM_011526495.2 | Intron | XP_011524797.1 | ||||
XM_011526496.2 | Intron | XP_011524798.1 | ||||
XM_011526497.2 | Intron | XP_011524799.1 | ||||
XM_017026333.1 | Intron | XP_016881822.1 |
LOC101928295 - uncharacterized LOC101928295 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |