Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGGGGTGGCCCATGGACCTGCC[C/T]GAGGGCCCGGTGGGTGGCCCCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC155 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC155 - coiled-coil domain containing 155 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144688.4 | 217 | Silent Mutation | CCC,CCT | P,P 4 | NP_653289.3 | |
XM_005258549.3 | 217 | Silent Mutation | CCC,CCT | P,P 4 | XP_005258606.1 | |
XM_011526488.2 | 217 | Silent Mutation | CCC,CCT | P,P 4 | XP_011524790.1 | |
XM_011526489.2 | 217 | Silent Mutation | CCC,CCT | P,P 4 | XP_011524791.1 | |
XM_011526490.2 | 217 | Silent Mutation | CCC,CCT | P,P 4 | XP_011524792.1 | |
XM_011526494.2 | 217 | UTR 5 | XP_011524796.1 | |||
XM_011526495.2 | 217 | UTR 5 | XP_011524797.1 | |||
XM_011526496.2 | 217 | Intron | XP_011524798.1 | |||
XM_011526497.2 | 217 | Silent Mutation | CCC,CCT | P,P 4 | XP_011524799.1 | |
XM_017026333.1 | 217 | Intron | XP_016881822.1 |
LOC101928295 - uncharacterized LOC101928295 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |