Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602464 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM222B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FAM222B - family with sequence similarity 222 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077498.2 | Intron | NP_001070966.1 | ||||
NM_001288631.1 | Intron | NP_001275560.1 | ||||
NM_001288632.1 | Intron | NP_001275561.1 | ||||
NM_001288633.1 | Intron | NP_001275562.1 | ||||
NM_001288634.1 | Intron | NP_001275563.1 | ||||
NM_001288635.1 | Intron | NP_001275564.1 | ||||
NM_001288636.1 | Intron | NP_001275565.1 | ||||
NM_001288637.1 | Intron | NP_001275566.1 | ||||
NM_001288638.1 | Intron | NP_001275567.1 | ||||
NM_001288639.1 | Intron | NP_001275568.1 | ||||
NM_001288640.1 | Intron | NP_001275569.1 | ||||
NM_018182.3 | Intron | NP_060652.2 | ||||
XM_011524986.2 | Intron | XP_011523288.1 | ||||
XM_011524987.1 | Intron | XP_011523289.1 | ||||
XM_011524988.2 | Intron | XP_011523290.1 | ||||
XM_017024831.1 | Intron | XP_016880320.1 | ||||
XM_017024832.1 | Intron | XP_016880321.1 | ||||
XM_017024833.1 | Intron | XP_016880322.1 | ||||
XM_017024834.1 | Intron | XP_016880323.1 | ||||
XM_017024835.1 | Intron | XP_016880324.1 |
TRAF4 - TNF receptor associated factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |