Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTAATTACACTACATTAAAAAT[C/T]ACCTAGTACTGAGCCAGATGTGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614275 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF565 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF565 - zinc finger protein 565 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042474.2 | 2373 | UTR 3 | NP_001035939.1 | |||
NM_152477.4 | 2373 | UTR 3 | NP_689690.3 | |||
XM_011526512.2 | 2373 | UTR 3 | XP_011524814.1 | |||
XM_011526514.2 | 2373 | UTR 3 | XP_011524816.1 | |||
XM_017026341.1 | 2373 | UTR 3 | XP_016881830.1 | |||
XM_017026342.1 | 2373 | UTR 3 | XP_016881831.1 | |||
XM_017026343.1 | 2373 | UTR 3 | XP_016881832.1 |
Set Membership: |
HapMap |