Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610518 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNTNAP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNTNAP4 - contactin associated protein like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322178.1 | Intron | NP_001309107.1 | ||||
NM_001322179.1 | Intron | NP_001309108.1 | ||||
NM_001322180.1 | Intron | NP_001309109.1 | ||||
NM_001322181.1 | Intron | NP_001309110.1 | ||||
NM_001322187.1 | Intron | NP_001309116.1 | ||||
NM_001322188.1 | Intron | NP_001309117.1 | ||||
NM_001322189.1 | Intron | NP_001309118.1 | ||||
NM_001322190.1 | Intron | NP_001309119.1 | ||||
NM_001322191.1 | Intron | NP_001309120.1 | ||||
NM_033401.4 | Intron | NP_207837.2 | ||||
NM_138994.4 | Intron | NP_620481.2 | ||||
XM_011523403.2 | Intron | XP_011521705.1 | ||||
XM_017023805.1 | Intron | XP_016879294.1 |