Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCGAGGTGGTGCTGACCTGGGG[G/A]ACTTGACACAGGCACGTGATCTTGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601065 MIM: 606490 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AARS PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AARS - alanyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001605.2 | 2825 | Silent Mutation | NP_001596.2 |
EXOSC6 - exosome component 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107984138 - serine/threonine-protein kinase SMG1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |