Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTGTGTGCTCTAGAGCTTTCTA[C/T]TGGATCAGAAGTTATCTCTAACTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 192090 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDH1 - cadherin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317184.1 | Intron | NP_001304113.1 | ||||
NM_001317185.1 | Intron | NP_001304114.1 | ||||
NM_001317186.1 | Intron | NP_001304115.1 | ||||
NM_004360.4 | Intron | NP_004351.1 | ||||
XM_011523488.1 | Intron | XP_011521790.1 | ||||
XM_011523489.1 | Intron | XP_011521791.1 |
Set Membership: |
HapMap |