Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614901 MIM: 609912 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCKDK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BCKDK - branched chain ketoacid dehydrogenase kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KAT8 - lysine acetyltransferase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032188.2 | Intron | NP_115564.2 | ||||
NM_182958.2 | Intron | NP_892003.2 | ||||
XM_011545970.1 | Intron | XP_011544272.1 | ||||
XM_011545971.1 | Intron | XP_011544273.1 |
Set Membership: |
HapMap |