Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616585 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM192A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FAM192A - family with sequence similarity 192 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RSPRY1 - ring finger and SPRY domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305163.1 | Intron | NP_001292092.1 | ||||
NM_001305164.1 | Intron | NP_001292093.1 | ||||
NM_001305182.1 | Intron | NP_001292111.1 | ||||
NM_133368.2 | Intron | NP_588609.1 | ||||
XM_005256220.1 | Intron | XP_005256277.1 | ||||
XM_011523427.1 | Intron | XP_011521729.1 | ||||
XM_011523428.1 | Intron | XP_011521730.1 | ||||
XM_011523430.1 | Intron | XP_011521732.1 | ||||
XM_017023844.1 | Intron | XP_016879333.1 |