Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATTTTTGTTATATACCGTTCACC[G/T]GTTCTGTAGTTTGTCAGGCAAGGTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604167 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTCF PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CTCF - CCCTC-binding factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191022.1 | Intron | NP_001177951.1 | ||||
NM_006565.3 | Intron | NP_006556.1 | ||||
XM_005255775.3 | Intron | XP_005255832.1 | ||||
XM_017022868.1 | Intron | XP_016878357.1 | ||||
XM_017022869.1 | Intron | XP_016878358.1 |
Set Membership: |
HapMap |