Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGCCAGTAAGCATTGGCTTAACT[G/T]TATATCCTTTCACGTACCAACTGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604917 MIM: 614463 MIM: 616424 | ||||||||||||||||||||
Literature Links: |
CNOT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNOT1 - CCR4-NOT transcription complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001265612.1 | Intron | NP_001252541.1 | ||||
NM_016284.4 | Intron | NP_057368.3 | ||||
NM_206999.2 | Intron | NP_996882.1 |
NDRG4 - NDRG family member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SETD6 - SET domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160305.2 | Intron | NP_001153777.1 | ||||
NM_024860.3 | Intron | NP_079136.2 | ||||
XM_011523336.1 | Intron | XP_011521638.1 |