Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610432 MIM: 616319 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF125 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RNF125 - ring finger protein 125 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF138 - ring finger protein 138 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191324.1 | Intron | NP_001178253.2 | ||||
NM_016271.4 | Intron | NP_057355.2 | ||||
NM_198128.2 | Intron | NP_937761.1 | ||||
XM_005258285.1 | Intron | XP_005258342.1 | ||||
XM_005258286.1 | Intron | XP_005258343.1 |
Set Membership: |
HapMap |