Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAAATTGAGAACAAAAGAATTTC[A/G]TAGGTTTTAATTTTTTAACCAAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600378 MIM: 611841 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IMMT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IMMT - inner membrane mitochondrial protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4779 - microRNA 4779 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL35 - mitochondrial ribosomal protein L35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016622.3 | Intron | NP_057706.2 | ||||
NM_145644.2 | Intron | NP_663619.1 | ||||
XM_005264361.2 | Intron | XP_005264418.1 |
Set Membership: |
HapMap |