Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGCCAAAGAAATTCACATATGGC[A/G]CATGATGCAGAAACAGTGAGAAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604911 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNOT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNOT4 - CCR4-NOT transcription complex subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008225.2 | Intron | NP_001008226.1 | ||||
NM_001190847.1 | Intron | NP_001177776.1 | ||||
NM_001190848.1 | Intron | NP_001177777.1 | ||||
NM_001190849.1 | Intron | NP_001177778.1 | ||||
NM_001190850.1 | Intron | NP_001177779.1 | ||||
NM_013316.3 | Intron | NP_037448.2 | ||||
XM_017012235.1 | Intron | XP_016867724.1 |