Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGGCTGTTTTCTGAGTTCAGACC[C/T]GGTGTGCTCTGCTTTGCATCCGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608757 MIM: 609725 MIM: 614586 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CLP1 - cleavage and polyadenylation factor I subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142597.1 | Intron | NP_001136069.1 | ||||
NM_006831.2 | Intron | NP_006822.1 |
YPEL4 - yippee like 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZDHHC5 - zinc finger DHHC-type containing 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |