Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612064 MIM: 139390 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GIGYF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GIGYF1 - GRB10 interacting GYF protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022574.4 | 4958 | UTR 3 | NP_072096.2 | |||
XM_005250532.1 | 4958 | UTR 3 | XP_005250589.1 | |||
XM_011516472.1 | 4958 | UTR 3 | XP_011514774.1 | |||
XM_011516477.1 | 4958 | UTR 3 | XP_011514779.1 | |||
XM_017012526.1 | 4958 | UTR 3 | XP_016868015.1 | |||
XM_017012527.1 | 4958 | UTR 3 | XP_016868016.1 | |||
XM_017012528.1 | 4958 | UTR 3 | XP_016868017.1 | |||
XM_017012529.1 | 4958 | UTR 3 | XP_016868018.1 | |||
XM_017012530.1 | 4958 | UTR 3 | XP_016868019.1 | |||
XM_017012531.1 | 4958 | UTR 3 | XP_016868020.1 |
GNB2 - G protein subunit beta 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |