Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 174760 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
POLB - DNA polymerase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002690.2 | Intron | NP_002681.1 | ||||
XM_005273535.3 | Intron | XP_005273592.1 | ||||
XM_005273536.3 | Intron | XP_005273593.1 | ||||
XM_005273537.3 | Intron | XP_005273594.1 | ||||
XM_005273538.2 | Intron | XP_005273595.1 | ||||
XM_005273539.2 | Intron | XP_005273596.1 | ||||
XM_005273540.4 | Intron | XP_005273597.1 | ||||
XM_006716353.2 | Intron | XP_006716416.1 | ||||
XM_017013583.1 | Intron | XP_016869072.1 | ||||
XM_017013584.1 | Intron | XP_016869073.1 |
Set Membership: |
HapMap |