Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTCATTTGGCTAATTATTATTA[A/C]AGTATTTAGTCTGATACCCTTCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP54 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CFAP54 - cilia and flagella associated protein 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306084.1 | 28 | Intron | NP_001293013.1 | |||
XM_011539069.2 | 28 | Intron | XP_011537371.1 | |||
XM_011539070.2 | 28 | Intron | XP_011537372.1 | |||
XM_011539071.2 | 28 | Intron | XP_011537373.1 | |||
XM_011539072.2 | 28 | Intron | XP_011537374.1 | |||
XM_011539073.2 | 28 | Intron | XP_011537375.1 | |||
XM_011539074.2 | 28 | Intron | XP_011537376.1 | |||
XM_011539076.2 | 28 | Intron | XP_011537378.1 | |||
XM_011539077.2 | 28 | UTR 5 | XP_011537379.1 | |||
XM_011539078.2 | 28 | Intron | XP_011537380.1 | |||
XM_011539079.1 | 28 | Intron | XP_011537381.1 | |||
XM_011539081.2 | 28 | Intron | XP_011537383.1 | |||
XM_011539083.2 | 28 | Intron | XP_011537385.1 | |||
XM_011539084.2 | 28 | Intron | XP_011537386.1 | |||
XM_011539085.1 | 28 | Intron | XP_011537387.1 | |||
XM_017018866.1 | 28 | Intron | XP_016874355.1 | |||
XM_017018867.1 | 28 | Intron | XP_016874356.1 |
Set Membership: |
HapMap |