Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605269 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CORO1C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CORO1C - coronin 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105237.2 | 2943 | UTR 3 | NP_001098707.1 | |||
NM_001276471.1 | 2943 | UTR 3 | NP_001263400.1 | |||
NM_014325.3 | 2943 | UTR 3 | NP_055140.1 | |||
XM_011538125.1 | 2943 | UTR 3 | XP_011536427.1 | |||
XM_017019121.1 | 2943 | UTR 3 | XP_016874610.1 | |||
XM_017019122.1 | 2943 | UTR 3 | XP_016874611.1 | |||
XM_017019123.1 | 2943 | UTR 3 | XP_016874612.1 |
Set Membership: |
HapMap |