Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTTTCCTCCGTCTAGGGGGGAT[C/T]CTAGCATCTGCCCTTCCATTTTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616598 MIM: 607175 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BORCS5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BORCS5 - BLOC-1 related complex subunit 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DUSP16 - dual specificity phosphatase 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030640.2 | 2714 | UTR 3 | NP_085143.1 | |||
XM_005253488.2 | 2714 | UTR 3 | XP_005253545.1 | |||
XM_006719155.2 | 2714 | UTR 3 | XP_006719218.1 | |||
XM_011520856.1 | 2714 | UTR 3 | XP_011519158.1 | |||
XM_011520857.1 | 2714 | UTR 3 | XP_011519159.1 | |||
XM_017019988.1 | 2714 | UTR 3 | XP_016875477.1 |
LOC105369662 - uncharacterized LOC105369662 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |