Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTGATGGGACGATTTTGCTGTC[A/G]TGTTTCTTACAAGGCAGATATAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613644 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATF7IP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATF7IP - activating transcription factor 7 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286514.1 | Intron | NP_001273443.1 | ||||
NM_001286515.1 | Intron | NP_001273444.1 | ||||
NM_018179.4 | Intron | NP_060649.3 | ||||
NM_181352.1 | Intron | NP_851997.1 | ||||
XM_005253424.3 | Intron | XP_005253481.1 | ||||
XM_006719108.3 | Intron | XP_006719171.1 | ||||
XM_006719109.3 | Intron | XP_006719172.1 | ||||
XM_011520754.2 | Intron | XP_011519056.1 | ||||
XM_011520755.2 | Intron | XP_011519057.1 | ||||
XM_011520756.2 | Intron | XP_011519058.1 | ||||
XM_017019638.1 | Intron | XP_016875127.1 | ||||
XM_017019639.1 | Intron | XP_016875128.1 |
Set Membership: |
HapMap |