Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102595 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACYP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACYP2 - acylphosphatase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320586.1 | Intron | NP_001307515.1 | ||||
NM_001320587.1 | Intron | NP_001307516.1 | ||||
NM_001320588.1 | Intron | NP_001307517.1 | ||||
NM_001320589.1 | Intron | NP_001307518.1 | ||||
NM_001320590.1 | Intron | NP_001307519.1 | ||||
NM_138448.3 | Intron | NP_612457.1 | ||||
XM_017005406.1 | Intron | XP_016860895.1 | ||||
XM_017005407.1 | Intron | XP_016860896.1 | ||||
XM_017005408.1 | Intron | XP_016860897.1 | ||||
XM_017005409.1 | Intron | XP_016860898.1 | ||||
XM_017005410.1 | Intron | XP_016860899.1 | ||||
XM_017005411.1 | Intron | XP_016860900.1 | ||||
XM_017005412.1 | Intron | XP_016860901.1 | ||||
XM_017005413.1 | Intron | XP_016860902.1 |
Set Membership: |
HapMap |