Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600523 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIGD1C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIGD1C - HIG1 hypoxia inducible domain family member 1C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC11A2 - solute carrier family 11 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000617.2 | 2615 | Intron | NP_000608.1 | |||
NM_001174125.1 | 2615 | Intron | NP_001167596.1 | |||
NM_001174126.1 | 2615 | UTR 3 | NP_001167597.1 | |||
NM_001174127.1 | 2615 | UTR 3 | NP_001167598.1 | |||
NM_001174128.1 | 2615 | Intron | NP_001167599.1 | |||
NM_001174129.1 | 2615 | Intron | NP_001167600.1 | |||
NM_001174130.1 | 2615 | Intron | NP_001167601.1 | |||
XM_005268911.3 | 2615 | UTR 3 | XP_005268968.1 | |||
XM_005268912.4 | 2615 | UTR 3 | XP_005268969.1 | |||
XM_011538404.2 | 2615 | UTR 3 | XP_011536706.1 | |||
XM_011538405.2 | 2615 | UTR 3 | XP_011536707.1 | |||
XM_017019355.1 | 2615 | UTR 3 | XP_016874844.1 | |||
XM_017019356.1 | 2615 | UTR 3 | XP_016874845.1 |