Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTTCAAATGAGATAATCCATAT[C/G]AAAAATTCAGCACTTTGTCTGACAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601439 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCC9 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ABCC9 - ATP binding cassette subfamily C member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005691.3 | Intron | NP_005682.2 | ||||
NM_020297.3 | Intron | NP_064693.2 | ||||
XM_005253284.3 | Intron | XP_005253341.1 | ||||
XM_005253286.3 | Intron | XP_005253343.1 | ||||
XM_005253287.4 | Intron | XP_005253344.1 | ||||
XM_005253288.3 | Intron | XP_005253345.1 | ||||
XM_005253289.3 | Intron | XP_005253346.1 | ||||
XM_005253290.3 | Intron | XP_005253347.1 | ||||
XM_006719025.3 | Intron | XP_006719088.1 | ||||
XM_011520545.2 | Intron | XP_011518847.1 |
Set Membership: |
HapMap |