Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTACTGACTGGTCCTTTCCACACC[C/T]CATCCTGGCTGCCAGTGGGCTTCAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603996 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF114 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF114 - zinc finger protein 114 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301062.1 | Intron | NP_001287991.1 | ||||
NM_153608.2 | Intron | NP_705836.1 | ||||
XM_011526574.2 | Intron | XP_011524876.1 | ||||
XM_017026414.1 | Intron | XP_016881903.1 | ||||
XM_017026415.1 | Intron | XP_016881904.1 |
Set Membership: |
HapMap |