Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612331 MIM: 603144 MIM: 180740 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf73 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C19orf73 - chromosome 19 open reading frame 73 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIN7B - lin-7 homolog B, crumbs cell polarity complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308419.1 | Intron | NP_001295348.1 | ||||
NM_022165.2 | Intron | NP_071448.1 | ||||
XM_006723323.3 | Intron | XP_006723386.1 | ||||
XM_017027131.1 | Intron | XP_016882620.1 |
PPFIA3 - PTPRF interacting protein alpha 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNRNP70 - small nuclear ribonucleoprotein U1 subunit 70 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |