Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTCTTACCAAGTAAGATTTCCC[A/G]TTTGAACCCCATCTCCCAGATACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608826 MIM: 176268 MIM: 162662 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CGB7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CGB7 - chorionic gonadotropin beta subunit 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCNA7 - potassium voltage-gated channel subfamily A member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NTF4 - neurotrophin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006179.4 | Intron | NP_006170.1 | ||||
XM_005258962.3 | Intron | XP_005259019.1 | ||||
XM_006723232.3 | Intron | XP_006723295.1 | ||||
XM_011527008.2 | Intron | XP_011525310.1 | ||||
XM_011527009.2 | Intron | XP_011525311.1 | ||||
XM_011527010.2 | Intron | XP_011525312.1 |
Set Membership: |
HapMap Validated |