Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGAGGAGAATCAGGGGAGGCTG[A/G]GAAGGGAATTTCTGGGTCTGGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604638 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTN4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ACTN4 - actinin alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322033.1 | Intron | NP_001308962.1 | ||||
NM_004924.5 | Intron | NP_004915.2 | ||||
XM_005259281.4 | Intron | XP_005259338.1 | ||||
XM_006723406.2 | Intron | XP_006723469.1 | ||||
XM_017027331.1 | Intron | XP_016882820.1 |