Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAGATGCTGCAGGGTGCCACCC[C/T]TGAGTCCCACAGTTAGGGTCCTTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615470 MIM: 604144 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP89 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CEP89 - centrosomal protein 89 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032816.4 | 4205 | Intron | NP_116205.3 | |||
XM_005259344.3 | 4205 | UTR 3 | XP_005259401.1 | |||
XM_011527425.2 | 4205 | UTR 3 | XP_011525727.1 | |||
XM_017027398.1 | 4205 | Intron | XP_016882887.1 |
SLC7A9 - solute carrier family 7 member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |