Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142705 MIM: 603144 MIM: 606936 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HRC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HRC - histidine rich calcium binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPFIA3 - PTPRF interacting protein alpha 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRPM4 - transient receptor potential cation channel subfamily M member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195227.1 | 115 | UTR 5 | NP_001182156.1 | |||
NM_001321281.1 | 115 | UTR 5 | NP_001308210.1 | |||
NM_001321282.1 | 115 | UTR 5 | NP_001308211.1 | |||
NM_001321283.1 | 115 | UTR 5 | NP_001308212.1 | |||
NM_001321285.1 | 115 | UTR 5 | NP_001308214.1 | |||
NM_017636.3 | 115 | UTR 5 | NP_060106.2 | |||
XM_005259017.1 | 115 | Intron | XP_005259074.1 |
Set Membership: |
HapMap |