Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608414 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLCE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLCE1 - phospholipase C epsilon 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165979.2 | Intron | NP_001159451.1 | ||||
NM_001288989.1 | Intron | NP_001275918.1 | ||||
NM_016341.3 | Intron | NP_057425.3 | ||||
XM_006717885.3 | Intron | XP_006717948.1 | ||||
XM_006717888.3 | Intron | XP_006717951.1 | ||||
XM_006717889.3 | Intron | XP_006717952.1 | ||||
XM_006717890.2 | Intron | XP_006717953.1 | ||||
XM_011539849.2 | Intron | XP_011538151.1 | ||||
XM_011539850.2 | Intron | XP_011538152.1 | ||||
XM_011539851.2 | Intron | XP_011538153.1 | ||||
XM_011539852.2 | Intron | XP_011538154.1 | ||||
XM_017016310.1 | Intron | XP_016871799.1 | ||||
XM_017016311.1 | Intron | XP_016871800.1 | ||||
XM_017016312.1 | Intron | XP_016871801.1 |