Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTTCCTACTGTGTCATTGTGCTC[A/C]TGGAGAGAACTTGACGGAATATCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608143 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPAT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPAT4 - glycerol-3-phosphate acyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178819.3 | Intron | NP_848934.1 | ||||
XM_011544392.2 | Intron | XP_011542694.1 | ||||
XM_011544393.1 | Intron | XP_011542695.1 | ||||
XM_011544394.1 | Intron | XP_011542696.1 | ||||
XM_017013042.1 | Intron | XP_016868531.1 | ||||
XM_017013043.1 | Intron | XP_016868532.1 |
LOC102723729 - uncharacterized LOC102723729 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |