Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCCTAAAGCTGAAATAAGCTAC[A/G]TTTATAATCCAGAGCCTTAAAGTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179615 MIM: 602355 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAG1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RAG1 - recombination activating gene 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000448.2 | Intron | NP_000439.1 | ||||
XM_005253041.4 | Intron | XP_005253098.1 | ||||
XM_011520250.2 | Intron | XP_011518552.1 |
TRAF6 - TNF receptor associated factor 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |