Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTACCAGTAAAACTAAAAGTGCCT[A/G]CTTTTAGTTTTCACTGTATTTTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 169800 | ||||||||||||||||||||
Literature Links: |
CNDP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNDP2 - CNDP dipeptidase 2 (metallopeptidase M20 family) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168499.1 | 43 | UTR 5 | NP_001161971.1 | |||
NM_018235.2 | 43 | Intron | NP_060705.2 | |||
XM_005266728.2 | 43 | Intron | XP_005266785.1 | |||
XM_006722503.2 | 43 | Intron | XP_006722566.1 | |||
XM_011526071.2 | 43 | Intron | XP_011524373.1 | |||
XM_011526072.2 | 43 | Intron | XP_011524374.1 | |||
XM_011526073.2 | 43 | Intron | XP_011524375.1 |