Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615210 MIM: 615423 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCNP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PCNP - PEST proteolytic signal containing nuclear protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320395.1 | Intron | NP_001307324.1 | ||||
NM_001320397.1 | Intron | NP_001307326.1 | ||||
NM_001320398.1 | Intron | NP_001307327.1 | ||||
NM_001320399.1 | Intron | NP_001307328.1 | ||||
NM_001320400.1 | Intron | NP_001307329.1 | ||||
NM_001320401.1 | Intron | NP_001307330.1 | ||||
NM_020357.2 | Intron | NP_065090.1 | ||||
XM_017006922.1 | Intron | XP_016862411.1 |
TRMT10C - tRNA methyltransferase 10C, mitochondrial RNase P subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |