Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGATAATTTTATTATGATGCAC[A/G]AGAACAGAAACATATCAGAATGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601529 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NR2C1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NR2C1 - nuclear receptor subfamily 2 group C member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032287.2 | Intron | NP_001027458.1 | ||||
NM_001127362.1 | Intron | NP_001120834.1 | ||||
NM_003297.3 | Intron | NP_003288.2 | ||||
XM_005269133.2 | Intron | XP_005269190.1 | ||||
XM_006719584.2 | Intron | XP_006719647.1 | ||||
XM_006719585.2 | Intron | XP_006719648.1 | ||||
XM_011538716.2 | Intron | XP_011537018.1 | ||||
XM_011538717.2 | Intron | XP_011537019.1 |