Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612449 MIM: 609387 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GEN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GEN1 - GEN1, Holliday junction 5' flap endonuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130009.2 | Intron | NP_001123481.2 | ||||
NM_182625.4 | Intron | NP_872431.4 | ||||
XM_005262613.4 | Intron | XP_005262670.1 | ||||
XM_006712005.3 | Intron | XP_006712068.1 | ||||
XM_011532820.2 | Intron | XP_011531122.1 | ||||
XM_011532821.2 | Intron | XP_011531123.1 | ||||
XM_011532822.2 | Intron | XP_011531124.1 |
SMC6 - structural maintenance of chromosomes 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142286.1 | Intron | NP_001135758.1 | ||||
NM_024624.5 | Intron | NP_078900.1 | ||||
XM_011533107.2 | Intron | XP_011531409.1 | ||||
XM_011533108.2 | Intron | XP_011531410.1 | ||||
XM_017004913.1 | Intron | XP_016860402.1 | ||||
XM_017004914.1 | Intron | XP_016860403.1 | ||||
XM_017004915.1 | Intron | XP_016860404.1 | ||||
XM_017004916.1 | Intron | XP_016860405.1 |
Set Membership: |
HapMap |