Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605768 | ||||||||||||||||||||||||||||||||
Literature Links: |
DAPP1 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
DAPP1 - dual adaptor of phosphotyrosine and 3-phosphoinositides 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306151.1 | Intron | NP_001293080.1 | ||||
NM_014395.2 | Intron | NP_055210.2 | ||||
XM_011531840.2 | Intron | XP_011530142.1 | ||||
XM_011531842.1 | Intron | XP_011530144.1 | ||||
XM_011531843.2 | Intron | XP_011530145.1 | ||||
XM_017008023.1 | Intron | XP_016863512.1 | ||||
XM_017008024.1 | Intron | XP_016863513.1 |