Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGCCTTTGCTTACTAAACTGAA[C/T]GAGAGGATATGAAGTAAGTGACTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137192 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GABRB3 - gamma-aminobutyric acid type A receptor beta3 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000814.5 | 2157 | UTR 3 | NP_000805.1 | |||
NM_001191320.1 | 2157 | UTR 3 | NP_001178249.1 | |||
NM_001191321.2 | 2157 | UTR 3 | NP_001178250.1 | |||
NM_001278631.1 | 2157 | UTR 3 | NP_001265560.1 | |||
NM_021912.4 | 2157 | UTR 3 | NP_068712.1 | |||
XM_011521428.2 | 2157 | UTR 3 | XP_011519730.1 |
Set Membership: |
HapMap Validated |