Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BBX PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BBX - BBX, HMG-box containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142568.2 | Intron | NP_001136040.1 | ||||
NM_001276286.1 | Intron | NP_001263215.1 | ||||
NM_020235.6 | Intron | NP_064620.2 | ||||
XM_005247642.3 | Intron | XP_005247699.1 | ||||
XM_005247643.3 | Intron | XP_005247700.1 | ||||
XM_005247644.3 | Intron | XP_005247701.1 | ||||
XM_011512999.2 | Intron | XP_011511301.1 | ||||
XM_011513000.1 | Intron | XP_011511302.1 | ||||
XM_011513001.1 | Intron | XP_011511303.1 | ||||
XM_011513002.2 | Intron | XP_011511304.1 | ||||
XM_011513003.2 | Intron | XP_011511305.1 | ||||
XM_011513004.2 | Intron | XP_011511306.1 | ||||
XM_011513005.2 | Intron | XP_011511307.1 | ||||
XM_011513006.2 | Intron | XP_011511308.1 | ||||
XM_011513007.2 | Intron | XP_011511309.1 | ||||
XM_011513010.2 | Intron | XP_011511312.1 | ||||
XM_017006881.1 | Intron | XP_016862370.1 | ||||
XM_017006882.1 | Intron | XP_016862371.1 | ||||
XM_017006883.1 | Intron | XP_016862372.1 |
Set Membership: |
HapMap |